Order Entry
United States
ContactUsLinkComponent
 
Anti-NFASC Goat Polyclonal Antibody
  10114-414
 :  Prosci
undefined
Anti-NFASC Goat Polyclonal Antibody
  10114-414
 :  Prosci
 :  42-393
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    neurofascin
  • Antigen Symbol:
    NFASC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human,
    Rat
  • Western Blot:
    Yes
  • Size:
    0.1 mg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    O94856
  • Antigen Synonyms:
    NFASC
  • Storage Buffer:
    NFASC antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
  • Storage Temperature:
    Aliquot and store at –20 °C. Minimize freezing and thawing.
  • Concentration:
    500 ug/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    NFASC antibody was raised against a amino acid synthetic peptide near the internal region of NFASC (aa 877-890).
  • Purification:
    NFASC antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Cat. No.:
    10114-414
  • Supplier no.:
    42-393

 

 

ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 170 kDa band observed in human lung and rat brain lysates (calculated MW of 138 kDa according to NP_001005388.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Kirschbaum et al, J Biol Chem. 2009 Oct 16;284 (42) :28533-42. PMID: 19666467). Recommended concentration: 0.5-2 ?g/mL.

NFASC antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NFASC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat