Order Entry
United States
ContactUsLinkComponent
Anti-UPF3A Goat Polyclonal Antibody
Anti-UPF3A Goat Polyclonal Antibody
Catalog # 10114-194
Supplier:  Prosci
undefined
Anti-UPF3A Goat Polyclonal Antibody
Catalog # 10114-194
Supplier:  Prosci
Supplier Number:  42-211
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    UPF3 regulator of nonsense transcripts homolog A (yeast)
  • Antigen Symbol:
    UPF3A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.1 mg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    Q9H1J1
  • Antigen Synonyms:
    UPF3A
  • Storage Buffer:
    UPF3A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
  • Storage Temperature:
    Aliquot and store at –20 °C. Minimize freezing and thawing.
  • Concentration:
    500 ug/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    UPF3A antibody was raised against a 13 amino acid synthetic peptide near the internal region of UPF3A.
  • Purification:
    UPF3A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Cat. No.:
    10114-194
  • Supplier no.:
    42-211

Specifications

About this item

ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 48 kDa band in lysates of cell lines HeLa and Kelly after 0.3 ?g/mL antibody staining. . The 48 kDa band was successfully blocked by incubation with the immunizing peptide.

UPF3A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: UPF3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: