Anti-ANKRD11 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
ANKRD11 is a member of a novel family of ankyrin repeats containing cofactors (ANCOs) that interact with p160 coactivators to inhibit ligand-dependent transactivation. ANKRD11 encodes a large nuclear protein with five ankyrin repeats, and parts of its sequences have been reported as nasopharyngeal carcinoma susceptibility protein and medulloblastoma antigen. This gene also colocalizes and interacts with histone deacetylases.
ANKRD11 antibody can be used for detection of ANKRD11 by ELISA at 1:62500. ANKRD11 antibody can be used for detection of ANKRD11 by western blot at 1.25 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: ANKRD11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog
Documents
10105-636.pdf
Datasheet
Specifications
- Size:100 ul
- Clonality:Polyclonal
- Molecular Weight:298 kDa
- Purification:Antibody is purified by protein A chromatography method.
- Form:Liquid
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Host:Rabbit
- Cat. No.:10105-636
- Reactivity:Human|Dog|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:4 °C
- Antibody Type:Primary
- Antigen Symbol:ANKRD11
- Gene ID:Q9UHR3
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human ANKRD11.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:ANKRD11
- Antigen Name:ankyrin repeat domain 11
- ELISA:Yes
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store ANKRD11 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.



