To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria
- Antigen Symbol:MMADHC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9H3L0
- Antigen Synonyms:MMADHC
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:33 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store C2orf25 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human C2orf25.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10104-316
- Supplier no.:26-997
Specifications
About this item
The function of C2orf25 remains unknown.Vitamin B12 (cobalamin) is an essential cofactor in several metabolic pathways. Intracellular conversion of cobalamin to adenosylcobalamin in mitochondria and to methylcobalamin in cytoplasm is necessary for homeostasis of methylmalonic acid and homocysteine. C2ORF25 encodes a protein involved in an early step of cobalamin metabolism (Coelho et al., 2008 [PubMed 18385497]).
C2orf25 antibody can be used for detection of C2orf25 by ELISA at 1:62500. C2orf25 antibody can be used for detection of C2orf25 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: MMADHC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat