Order Entry
United States
ContactUsLinkComponent
Anti-RFP Chicken Polyclonal Antibody
Anti-RFP Chicken Polyclonal Antibody
Certificates
undefined
Anti-RFP Chicken Polyclonal Antibody
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • RL600-901-379
  • RL600-901-379S
  • Antigen Symbol:
  • RFP
  • RFP
  • Antigen Name:
  • Red Fluorescent Protein
  • Red Fluorescent Protein
  • Antigen Synonyms:
  • DSRED
  • DSRED
  • Antibody Type:
  • Tag
  • Tag
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Chicken
  • Chicken
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Epitope:
  • Full length
  • Full length
  • Immunogen:
  • The immunogen is a Red Fluorescent Protein (RFP) fusion Protein corresponding to the full length amino acid sequence (234aa) derived from the mushroom polyp coral Discosoma.
  • Red Fluorescent Protein (RFP) fusion Protein corresponding to the full length amino acid sequence (234aa) derived from the mushroom polyp coral Discosoma.
  • Format:
  • Antibody fraction
  • Antibody fraction
  • Form:
  • Liquid (sterile filtered)
  • Liquid (sterile filtered)
  • Purification:
  • filtration, delipidation, salt fractionation and extensive dialysis against the buffer stated above
  • filtration, delipidation, salt fractionation and extensive dialysis against the buffer stated above
  • Concentration:
  • 1.0 mg/ml by UV absorbance at 280 nm
  • 1.0 mg/ml by UV absorbance at 280 nm
  • Cross Adsorption:
  • No
  • No
  • Storage Buffer:
  • 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
  • 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
  • Storage Temperature:
  • –20 °C
  • –20 °C
  • Shipping Temperature:
  • Dry Ice
  • Dry Ice

Specifications

Frequently Bought Together