To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Thyroid Hormone Receptor, Beta
- Antigen Symbol:THRB
- Clonality:Monoclonal
- Clone:2386
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Epitope:N-Terminal
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:7068
- Antigen Synonyms:GRTH|ERBA-BETA|THRB2|NR1A2|ERBA2|MGC126109|thyroid hormone receptor|avian)|oncogene ERBA2|THRB1|beta|MGC126110|THR1|thyroid hormone receptor beta 1|avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2|generalized resistance to thyroid hormone|beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2|Thyroid hormone receptor
- Storage Buffer:0.01 M HEPES, 0.15 M Sodium Chloride, pH 7.50.1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage Temperature:–20 °C
- Shipping Temperature:Dry Ice
- Immunogen:Anti-Thyroid Hormone Receptor beta monoclonal antibody was produced in mouse by repeated immunizations with synthetic peptide corresponding to amino acid residues from the N-terminal region of Thyroid Hormone Receptor beta.
- Purification:Protein G purified
- Cat. No.:RL209-301-A96
- Supplier no.:209-301-A96
Anti-Thyroid Hormone Receptor beta (Mouse) antibody is suitable for use in Western Blotting. Expect a band of approximately 55 kDa corresponding to TR--beta Proteins in the appropriate cell lysate or extract.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: THRB
Clonality: Monoclonal
Clone: 2386
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human