To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Orosomucoid 1
- Antigen Symbol:ORM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:25 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Antigen Synonyms:Orosomucoid 1|agpa|ORM1|Alpha 1 agp|orm|Alpha 1 acid glycoprotein|omd1|Glycoprotein alpha acid of serum|OMD 1
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide
- Storage Temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:Anti-Alpha-1-Acid Glycoprotein Antibody was produced by repeated immunizations with human plasma a-1-Acid Glycoprotein.
- Purification:purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Cat. No.:RL200401046S
- Supplier no.:200-401-046S
Specifications
About this item
Anti-Alpha-1-Acid Glycoprotein Antibody is suitable for western blottingand for ELISA. Researchers should determine optimal titers for applications that are not stated below.
Recommended Dilutions: ELISA: 1:135000; Western Blot: 1:2000 - 1:10000; Immunochemistry: User Optimized; Other Dilution: User Optimized
Type: Primary
Antigen: ORM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human