PCR Tube Labels, Brady
: Avantor
Designed for use in labs, this material has a very low-profile construction that prevents vials from jamming in the centrifuge or racks. It stays put in the harshest conditions, and the clear tail can be wrapped over text for added protection from solvents. For use in liquid nitrogen, label must wrap around vial and slightly overlap itself.
- Liquid nitrogen (–196 °C)
- Hot water bath (100 °C)
- Freezer (–80 °C)
- Autoclave (121 °C)
- Sticks to frozen/frosted surfaces
FreezerBondz Material:
If you ever need to put a label on an already-frozen vial, this material will adhere to a frozen or frosted surface without the need to scrape or dry the vial before placement of the label. To be assured that the label stays secured when placed back into storage it is recommended that the label wrap around the vial and slightly overlap itself.
Frequently Bought Together