ContactUsLinkComponent
Pipettor Workstation, Heathrow Scientific
  490007-634
undefined
Pipettor Workstation, Heathrow Scientific
  490007-634
  • Description:
    Pipettor Workstation
  • Environmentally Preferable:
  • Cat. No.:
    490007-634
  • Supplier no.:
    HS234631
Organize your pipettors and tips
Acrylic rack holds up to four pipettors on top shelf; dual bins house two different sizes of tips. 244 x 165 x 246 mm.