- Description:SEM Paper Storage Box
- Environmentally Preferable:
- Cat. No.:102091-316
- Supplier no.:76505
The pop-up box is an inexpensive storage solution for storing scanning electron microscope pin mounts.The boxh stores up to 8 mounts: 1/2" (12.7 mm) head, 1/8" (3.2 mm) diameter pin. Box dimensions: 7 cm long×3.8 cm wide×3.5 cm high.