ContactUsLinkComponent
Sapphire Blade/Knife, Electron Microscopy Sciences
Sapphire Blade/Knife, Electron Microscopy Sciences
Catalog # 100492-502
Minority or Woman-Owned Business Enterprise
undefined
Sapphire Blade/Knife, Electron Microscopy Sciences
Catalog # 100492-502
Supplier Number:  93060
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Sapphire Knife
  • Environmentally Preferable:
  • Cat. No.:
    100492-502
  • Supplier no.:
    93060
Specifications
About this item
A unique high-precision knife for use with all Vibratomes and Tissue Sectioners, as well as Oscillating Tissue Slicers (EMS 4000 and EMS 5000).
This Sapphire Knife offers smooth, consistent and thinner sections than are usually obtainable with stainless steel blades. The blade allows for sectioning as thin as 10 microns. Offering many years of cutting surface, the durability of our sapphire knife far outlasts other comparable blades on the market.
Material: Synthetic Single Crystal Sapphire
Size: 1.5 x 0.4 x .03”