To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Petri Dish Incubation Tray
- Environmentally Preferable:
- Cat. No.:100489-262
- Supplier no.:71395-03
Specifications
About this item
Trays for the organization of all of your Petri dishes. They are designed to hold five 10 cm diameter or fifteen 6 cm diameter Petri dishes. Each tray comes with two large label areas for critical sample identification by either using a marker or a label.
- Increases incubator capacity
- Stackable
- Transport and store Petri dishes with ease while maintaining even CO2 circulation
- Easy sample grouping and identification
- Made from polypropylene and autoclavable
- Tray measures 97/8 x 95/16 x 13/8" high (251 x 237 x 35 mm)