About this item
LR White is a polar monomer polyhydroxylated acromatic acrylic resin. It has a very low viscosity (8cps), is non-toxic resin, and is suitable for both Light and Electron Microscopy.
- Can be cured by heat or by UV light
- Polymerized LR White resin are hydrophilic
- Available in medium and hard grade
Sections of polymerized LR White resin are hydrophilic. This character allows immuno-cytochemistry reagents to easily penetrate into the sections and etching of the sections is not necessary (etching sections will effect delicate tissue antigens). LR White sections also show minimal non-specific staining. Its low viscosity makes it ideal for infiltrating decalcified bone and teeth as well as plant tissues.