About this item
Human peripheral blood mononuclear cells (PBMC) are collected from healthy human volunteer donors following cGMP and cGTP collection guidelines under IRB approved informed consent by leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol
Mononuclear cells are widely used in research and clinical applications such as in microbiology, virology, oncology, vaccine development, transplant and regenerative biology, and toxicology.
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugal technology directly into the collection bag. Mononuclear cells are purified using density gradient centrifugation techniques.