Order Entry
United States
ContactUsLinkComponent
Di-tert-butyl peroxide for synthesis, Sigma-Aldrich®
Di-tert-butyl peroxide for synthesis, Sigma-Aldrich®
Catalog # EM8.20248.0250
Supplier:  MilliporeSigma
CAS Number:  
Di-tert-butyl peroxide for synthesis, Sigma-Aldrich®
Catalog # EM8.20248.0250
Supplier:  MilliporeSigma
Supplier Number:  8.20248.0250
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    250 ml
  • Environmentally Preferable:
  • Cat. No.:
    EM8.20248.0250
  • Supplier no.:
    8.20248.0250

Specifications

Specification Test Results

  • Solubility:
    0.063 g/l (20 °C)
  • Molar Mass:
    146.23 g/mol
  • Boiling Point:
    110 - 115 °C
  • Vapor Pressure:
    1000 hPa (110 °C)
  • Flash Point:
    6 °C
  • Density:
    0.80 g/cm3 (20 °C)
  • Ignition Point:
    182 °C

Specification Test Results

About this item

Synonyms: Luperox(R) DI, tert-Butyl peroxide, tert-Butyl peroxide
 :
 :146.23
UN:3107
 :GHS02
 :GHS08
CAS Number:110-05-4
Formula:C₈H₁₈O₂
MDL Number:MFCD00008803
Boiling Pt:109
Density:0.798
Flash Pt:12
ADR:5.2
Merck Index:12,03515
 :M-Clarity™ quality level = MQ200