To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:protein tyrosine phosphatase, receptor type, D
- Antigen Symbol:PTPRD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P23468
- Antigen Synonyms:PTPRD
- Storage Buffer:PTPRD Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:Predicted: 165, 210 kDa (130 kDa for cleavage product) Observed: 130 kDa
- Storage Temperature:PTPRD antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:PTPRD antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human PTPRD. The immunogen is located within the first 50 amino acids of PTPRD.
- Tested Applications:E, WB
- Purification:PTPRD Antibody is affinity chromatography purified via peptide column.
- Cat. No.:89418-184
- Supplier no.:6875
Specifications
About this item
PTPRD Antibody: PTPRD (Protein tyrosine phosphatase receptor type D) is a member of the protein tyrosine phosphatase (PTP) family that plays diverse roles during development including cell growth, differentiation, mitotic cycle and oncogenic transformation. PTPRD contains an extracellular region, a single transmembrane segment and two tandem intracytoplasmic catalytic domains. The extracellular region of PTPRD is composed of three Ig-like and eight fibronectin type III-like domains (1,3). PTPRD interacts with PPFIA1-3 and is a tumor suppressor on chromosome 9p that is involved in the development of glioblastoma multiforme (GBMs) and multiple human cancers.
Type: Primary
Antigen: PTPRD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human