To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:nucleolar protein 3 (apoptosis repressor with CARD domain)
- Antigen Symbol:NOL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O60936
- Antigen Synonyms:NOL3
- Storage Buffer:Nop30 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:Nop30 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping Temperature:4 °C
- Immunogen:Nop30 antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human Nop30. The immunogen is located within the last 50 amino acids of Nop30.
- Tested Applications:E, WB
- Purification:Nop30 Antibody is affinity chromatography purified via peptide column.
- Cat. No.:89415-378
- Supplier no.:2225
Specifications
About this item
Nop30 Antibody: Apoptosis, also known as programmed cell death, plays major roles in development and normal tissue turnover in addition to tumor formation. Apoptosis is regulated by death domain (DD) and/or caspase recruitment domain (CARD) containing molecules and the caspase family of proteases. CARD domain containing cell death regulators include RAIDD, Apaf-1, caspase-9, and caspase-2. A novel CARD domain containing protein was recently identified and designated ARC for apoptosis repressor with CARD. An alternate splicing isoform of ARC was identified as Nop30. While ARC interacts with caspase-2 and -8 and suppresses apoptosis induced by cell death adapters FADD and TRADD and by cell death receptors Fas, TNFR-1 and DR3, Nop30 multimerizes and binds to the splicing factor SRp30c and may act to influence alternative splice site selection in vivo. The Nop30 antibody will not detect ARC protein.
Type: Primary
Antigen: NOL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat