Order Entry
United States
ContactUsLinkComponent
Anti-GAPDH Rabbit Polyclonal Antibody
Anti-GAPDH Rabbit Polyclonal Antibody
Catalog # 89416-024
Supplier:  Prosci
undefined
Anti-GAPDH Rabbit Polyclonal Antibody
Catalog # 89416-024
Supplier:  Prosci
Supplier Number:  3783
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    glyceraldehyde-3-phosphate dehydrogenase
  • Antigen Symbol:
    GAPDH
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.1 mg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    P04406
  • Antigen Synonyms:
    GAPDH
  • Storage Buffer:
    GAPDH Antibody is supplied in PBS containing 0.02% sodium azide.
  • Storage Temperature:
    GAPDH antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    GAPDH antibody was raised against a 16 amino acid synthetic peptide from near the amino-terminus of human GAPDH. The immunogen is located within amino acids 60 - 110 of GAPDH.
  • Tested Applications:
    E, WB, IHC, IF
  • Purification:
    GAPDH Antibody is affinity chromatography purified via peptide column.
  • Cat. No.:
    89416-024
  • Supplier no.:
    3783

Specifications

About this item

GAPDH Antibody: Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) catalyzes the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD), an important energy-yielding step in carbohydrate metabolism. Recent evidence suggests that it also is involved in a number of cellular processes such as membrane fusion, phosphotransferase activity, DNA replication and repair, and nuclear RNA export. GAPDH has also been implicated in playing a role in different pathologies such as cancer progression, apoptosis, and neuronal diseases such as Alzheimer's and Huntington's disease. GAPDH is constitutively expressed at high levels in almost all tissues and cell lines making it ideal for use as a loading control marker in immunoblots.

Type: Primary
Antigen: GAPDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat