Order Entry
United States
ContactUsLinkComponent
Copper (Free and Total) Color Disc Test Kit, Model CU-6, Hach
Copper (Free and Total) Color Disc Test Kit, Model CU-6, Hach
Catalog # 89500-690
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
undefined
Copper (Free and Total) Color Disc Test Kit, Model CU-6, Hach
Catalog # 89500-690
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    Bicinchoninate
  • Environmentally Preferable:
  • Method:
    Color-Disc/Bicinchoninate, Hydrosulfite Reduction
  • Cat. No.:
    89500-690
  • Range:
    0 - 4 mg/L
  • Supplier no.:
    2194100
  • No. of tests:
    Free 50, total 50

Specifications

About this item

Copper may exist in natural waters and effluents as a soluble salt or as suspended solids

  • Simple, low-cost color disc method is the most accurate visual test kit method
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field

Hazardous Items. Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.

A small amount is essential for plants and animals, and concentrations exceeding 0.1 mg/L are useful for controlling algae and plankton growth. Quantities ranging from 0.02 to 0.1 mg/L are toxic for some fish, so its use for treating fish disease requires careful monitoring. This kit is USEPA-accepted for waste water and potable water and approved by some states for potable water under specific conditions, check with your regulatory agency. Contains 50 Free and 50 Total tests

Ordering information: Two reagents, color disc, comparator box, two viewing tubes, instructions and carrying case included.