Order Entry
United States
ContactUsLinkComponent
SpeI-HF®, Restriction Enzymes, New England Biolabs
SpeI-HF®, Restriction Enzymes, New England Biolabs
  102500-166
Small Business Enterprise
 :  
undefined
SpeI-HF®, Restriction Enzymes, New England Biolabs
  102500-166
 :  R3133M
Small Business Enterprise
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Concentration:
    100000 U/ml
  • Application:
  • Source:
  • Description:
    SpeI-HF®
  • Size:
    2500 units
  • Environmentally Preferable:
  • Enzyme Name:
    SpeI-HF
  • Cat. No.:
    102500-166
  • Supplier no.:
    R3133M

 

 

An E. coli strain that carries the cloned and modified SpeI gene from Sphaerotilus species (ATCC 13923). High Fidelity (HF®) Restriction Enzymes have 100% activity in CutSmart® Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver™ qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. Engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.