PH.D.™-12 Phage Display Peptide Library Kit, New England Biolabs
Catalog # 101227-568
Supplier: Avantor
Small Business Enterprise
CAS Number:
Specifications
- Description:Ph.D.-12 Phage Display Peptide Library Kit
- Size:10 exps
- Environmentally Preferable:
- Cat. No.:101227-568
- Supplier no.:E8110S
Specifications
About this item
The Ph.D.™-12 Phage Display Peptide Library is based on a combinatorial library of random dodecapeptides fused to a minor coat protein (pIII) of M13 phage
The displayed peptide (12-mer) is expressed at the N-terminus of pIII, i.e., the first residue of the mature protein is the first randomized position. The peptide is followed by a short spacer (Gly-Gly-Gly-Ser) and then the wild-type pIII sequence. The library consists of approximately 109 electroporated sequences amplified once to yield approximately 100 copies of each sequence in 10 µl of the supplied phage.