- Description:Fast DNA Ladder
- Size:50-200 gel lanes
- Environmentally Preferable:
- Cat. No.:101641-050
- Supplier no.:N3238S
The Fast DNA ladder is a pre-mixed, ready-to-load molecular weight marker containing xylene cyanol FF dye as a tracking dye
The DNA Ladder consists of proprietary plasmids, which are digested to completion with appropriate restriction enzymes to yield 11 bands suitable for use as molecular weight standards for fast electrophoresis systems as well as standard electrophoresis. The digested DNA includes fragments ranging from 50 base pairs to 10 kilobases. The 1 kb fragment has increased intensity to serve as reference band.