About this item
Capto™ Q is a strong anion exchange bioprocess medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective capture and intermediate protein purification when high volume throughput is essential.
- Strong anion exchanger for high-productivity capture and intermediate purification when high volume throughput is essential
- High dynamic binding capacity at high flow raises productivity
- High-volume throughput cuts process times
- Cost-effective processing with smaller unit operations
Capto Q withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime. Capto Q is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Capto Q meets industrial demands with security of supply and comprehensive technical and regulatory support.
Capto Q is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Capto Q meets industrial demands with security of supply and comprehensive technical and regulatory support.
Capto Q is designed for capture and intermediate purification of proteins from large feed volumes. It is composed of a rigid, high-flow agarose matrix modified with dextran surface extenders and a strong quaternary ammonium (Q) anion exchanger. The combination of high volume throughput and high dynamic binding capacity for Capto Q raises the productivity in industrial downstream processes. Capto Q withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime.
Specifications
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot Number17531602_INSTRUCTIONS 17531602_INSTRUCTIONS 1 17531602_INSTRUCTIONS 2 17531610_INSTRUCTIONS 17531610_INSTRUCTIONS 1