About this item
HiTrap Desalting are ready-to-use 5 ml columns prepacked with Sephadex G-25 Superfine resin for fast and convenient desalting and buffer exchange.
- Convenient desalting and buffer exchange of sample volumes up to 1.5 ml
- Two to five columns can be connected in series for sample sizes from 3 ml to 7.5 ml
- Replaces dialysis for desalting protein, which saves time and reduces labor
- Requires less than one minute with typically > 95% recovery
- HiTrap column format is convenient for use with a syringe, a peristaltic pump, or a chromatography system
HiTrap Desalting is a prepacked, ready-to-use column for group separation between high and low molecular weight substances. Applications include buffer exchange prior to or after different chromatographic steps, removal of low molecular weight contaminants, or removal of reagents to terminate a reaction.
The special design of the column, together with the well-known resin Sephadex G-25 Superfine, provides fast and reproducible separations in a convenient format.
The exclusion limit is approximately Mr 5000. This ensures group separation of proteins/peptides larger than Mr 5000 from smaller molecules.
Separations are easily performed with a syringe, a pump, or a chromatography system such as an ÄKTA system.
Specifications
- Catalog No:
- 95056-006
- 89501-384
- Max. Pressure:
- 5 bar [0.5 MPa] (70 psi)
- 5 bar [0.5 MPa] (70 psi)
- pH Range:
- 2 - 13
- 2 - 13
- Storage:
- 4...30 °C, 20% Ethanol
- 4...30 °C, 20% Ethanol
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements Licensing Statements GE Healthcare Life Sciences
Additional Product Details
GE Healthcare SEC Columns and Media Selection Guide