HisTrap Columns, FF Crude
Supplier: Cytiva
Certificates
About this item
HisTrap FF Crude columns are high binding capacity IMAC columns for his-tagged protein purification when scalability is needed.
- High binding capacity – binds up to 40 mg his-tag protein/mL resin
- Excellent flow properties – great performance even at high flow rates
- Optimized for unclarified samples – short purification time can minimize potential negative effects, such as degradation and oxidation of proteins
- HiTrap column format – compatible with syringe, pump, or chromatography system such as ÄKTA systems or other FPLC systems
HisTrap FF crude is a ready-to-use column, prepacked with precharged Ni Sepharose 6 Fast Flow. After thorough cell disruption, it is possible to load the unclarified lysate on the column, without precentrifugation and filtration.
Specifications
- Catalog No:
- 95055-840
- 89501-376
- 95056-136
- Description:
- HisTrap™ FF crude
- HisTrap™ FF crude
- HisTrap™ FF crude
- Binding Capacity:
- Approx. 40 mg histidine-tagged protein/ml medium
- Approx. 40 mg histidine-tagged protein/ml medium
- Approx. 40 mg histidine-tagged protein/ml medium
- Max. Pressure:
- 5 bar [0.5 MPa] (70 psi)
- 5 bar [0.5 MPa] (70 psi)
- 5 bar [0.5 MPa] (70 psi)
- pH Range:
- 3 - 12
- 3 - 12
- 3 - 12
- Storage:
- 4...30 °C, 20% Ethanol
- 4...30 °C, 20% Ethanol
- 4...30 °C, 20% Ethanol
Specifications
Related Information
Frequently Bought Together