Order Entry
United States
ContactUsLinkComponent
HiTrap Columns, Protein A HP, Cytiva
HiTrap Columns, Protein A HP, Cytiva
Supplier:  Cytiva
Certificates
undefined
HiTrap Columns, Protein A HP, Cytiva
Supplier:  Cytiva
Certificates

Specifications

  • Catalog No:
  • 89501-368
  • 95055-844
  • 95056-142
  • 95055-846
  • 95056-032
  • Max. Pressure:
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • pH Range:
  • 3 - 9
  • 3 - 9
  • 3 - 9
  • 3 - 9
  • 3 - 9
  • Storage:
  • 4 to 8 °C, 20% Ethanol
  • 4 to 8 °C, 20% Ethanol
  • 4 to 8 °C, 20% Ethanol
  • 4 to 8 °C, 20% Ethanol
  • 4 to 8 °C, 20% Ethanol

Specifications

Frequently Bought Together