PIG® HazMat Spill Kit in Extra-Large Response Chest, New Pig
Supplier: New Pig
Certificates
About this item
Seamless, polyethylene, lockable container keeps absorbents clean, dry and secure before use
- Prepacked chest provides easy access to contents
- Large, color-coded label makes identification fast and accurate
- Tamper-proof seals help prevent pilfering of spill response supplies
- Four-way forklift entry for easy moving
The 10" no flat wheels roll easily over uneven surfaces. The 5" rubber wheels are well suited to smooth concrete or pavement. Also available without wheels. Not UN rated.
PIG® HazMat Socks stop spreading spills; PIG® HazMat Pads and Pillows absorb; PIG® HazMat Pulp cleans up residue on rough surfaces. PIG® HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
Specifications
- Catalog No:
- 56620-216
- 56620-460
- 56620-462
- Capacity:
- Up to 408.8 L (108 gal.)
- Up to 408.8 L (108 gal.)
- Up to 408.8 L (108 gal.)