To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:Stearoyl-CoA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P13516
- Antigen Synonyms:Fatty Acid Desaturase 1|Acyl-CoA Desaturase 1|SCD1
- Storage Buffer:0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage Temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Synthetic peptide corresponding to aa 21-33 (A21PPSGNEREKVKT33) of mouse SCD1.
- Cat. No.:102514-460
- Supplier no.:AG25A0031C100
Specifications
About this item
Stearoyl-CoA desaturase (SCD) is a regulatory enzyme in lipogenesis. The enzyme is constitutively expressed in adipose tissue and is induced in liver in response to fat free, high carbohydrate diet. Several SCD isoforms (SCD1, -2, and -3) exist in the mouse. Mice with a targeted disruption of the SCD1 isoform had reduced body adiposity, increased insulin sensitivity, and resistance to diet-induced weight gain. The protection from obesity involved increased energy expenditure and increased oxygen consumption. SCD1 deficiency shows activation of lipid oxidation in addition to reduced triglyceride synthesis and storage.
Type:
Antigen: Stearoyl-CoA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse