To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:TNF Receptor Superfamily Member 10c
- Antigen Symbol:TRAIL-R3
- Clonality:Monoclonal
- Clone:TR3.06
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O14798
- Antigen Synonyms:TRAIL Receptor 3|TRID|DcR1|CD263|TNFRSF10C
- Storage Buffer:In PBS containing 10% glycerol and 0.02% sodium azide.
- Storage Temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Recombinant human TRAIL-R3 (DcR1).
- Tested Applications:FACS
- Cat. No.:102514-322
- Supplier no.:AG20B0029C100
Specifications
About this item
TRAIL-R3 is a receptor for the cytotoxic ligand TRAIL. It lacks a cytoplasmic death domain and hence is not capable of inducing apoptosis. Protects cells against TRAIL mediated apoptosis by competing with TRAIL-R1 and R2 for binding to the ligand.
- Stable for at least 1 year after receipt when stored at -20°C.
Purified from concentrated hybridoma tissue culture supernatant.
Purity: >95% (SDS-PAGE)
Type:
Antigen: TRAIL-R3
Clonality: Monoclonal
Clone: TR3.06
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human