Order Entry
United States
ContactUsLinkComponent
Anti-Cardif Mouse Monoclonal Antibody [clone: Adri-1]
Anti-Cardif Mouse Monoclonal Antibody [clone: Adri-1]
Catalog # 102514-260
Supplier:  Adipogen
Anti-Cardif Mouse Monoclonal Antibody [clone: Adri-1]
Catalog # 102514-260
Supplier:  Adipogen
Supplier Number:  AG-20B-0004-C100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    Cardif
  • Clonality:
    Monoclonal
  • Clone:
    Adri-1
  • Conjugation:
    Unconjugated
  • Host:
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    Q7Z434
  • Antigen Synonyms:
    CARD Adapter Inducing Interferon-beta|Virus-induced Signaling Adapter|MAVS|Interferon-beta Promoter Stimulator Protein 1|VISA|IPS-1|Mitochondrial Antiviral Signaling Protein
  • Storage Buffer:
    In PBS containing 10% glycerol and 0.02% sodium azide.
  • Storage Temperature:
    Short term: +4 °C, Long Term: –20 °C
  • Concentration:
    1mg/ml
  • Shipping Temperature:
    -20° C
  • Immunogen:
    Recombinant human Cardif (aa 160-450).
  • Cat. No.:
    102514-260
  • Supplier no.:
    AG20B0004C100

Specifications

About this item

RIG-I (retinoic acid-inducible gene I; Ddx58) and MDA5 (melanoma differentiation-associated gene 5, also known as Ifih1 or Helicard) are proteins that sense viral replication intermediates, such as double-stranded RNA and triggers the host antiviral programs. These molecules signal the downstream activation of NF-kappaB and IFN regulatory factor (IRF) -3, which coordinately regulate the expression of type-I interferons. Cardif (also called VISA/IPS-1/MAVS) is a new CARD (caspase activation and recruitment domain)-containing adaptor protein that interacts with the CARD domain of RIG-I and MDA5, leading to the activation of NF-kappaB and IRF3. Cardif is located to the mitochondrial outer membrane. Removal of the mitochondrial-targeting domain of cardif abolishes its ability to induce IFNs. Cardif is cleaved and inactivated by NS3-4A, a serine protease from hepatitis C virus known to block interferon-beta production.

  • Stable for at least 1 year after receipt when stored at -20°C.

Purified from concentrated hybridoma tissue culture supernatant.

Purity: >95% (SDS-PAGE)

Type:
Antigen: Cardif
Clonality: Monoclonal
Clone: Adri-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human