To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:
- Species:
- Size:5 mg
- Environmentally Preferable:
- Protein/Peptide Name:Z-AAF-CMK
- Purity:98%
- Cat. No.:89161-624
- Supplier no.:PI124-0005
Specifications
About this item
Negative control for AAF-CMK
Negative control for serine protease inhibitor AAF-CMK which inhibits tripeptidyl peptidase II (TPPII), a giant protease which may substitute for some proteasome functions. Blockage of the N-terminus by the Benzyloxycarbonyl or “Z” group abolishes the activity of the AAF-CMK inhibitor.