To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:
- Species:
- Size:5 mg
- Environmentally Preferable:
- Protein/Peptide Name:RGD Peptide
- Purity:>97% (HPLC)
- Cat. No.:89161-294
- Supplier no.:P700-0005
Specifications
About this item
Integrin ligand
Induces apoptosis in various cell types and cell lines at 1mM by a mechanism involving cellular uptake and possible direct stimulation of procaspase-3 autoprocessing. Does not induce apoptosis in the breast cancer line MCF-7, in which there is a functional deletion of the procaspase-3 gene. Contains the motif RGD (Arg-Gly-Asp) which is found in many integrin ligands. Inhibits cell adhesion of COS-7 cells to fibronectin 80-90% at 1mM.