To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Syntaxin 13
- Antigen Symbol:STX13
- Clonality:Monoclonal
- Host:Mouse
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
- Purification:Protein G affinity purified
- Cat. No.:200062-934
- Supplier no.:VAM-SV026E
Specifications
About this item
Syntaxins are type II integral membrane proteins localized to membranes that regulate vesicle transport, protein maturation, and exocytosis. Syntaxin 1A resides in the nerve terminals of sensory neurons and nerve fibers reaching small blood vessels. Syntaxin 2 localizes to the apical plasma membrane and regulates epithelial-mesenchymal interactions. Syntaxin 6 is primarily localized to the trans-Golgi network, and partially co-localizes with AP-1 on clathrin-coated membranes. Syntaxin 13 is ubiquitously expressed and localizes to the tubular early and recycling endosomes, as well as endosomal vacuoles.
- For Western Blot
Recognizes mouse, rat, dog and hamster Syntaxin 13. Detects a band of ~38kDa by Western blot.
Type: Primary
Antigen: STX13
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse