Hsp60 is a member of the chaperonin family of heat shock proteins, with homologs functioning in the cytosol and mitochondria to fold nascent and aggregated proteins. Hsp60 is the eukaryotic homolog of the E. coli GroEL protein, and forms a multimeric complex in the mitochondria with Hsp10 (Cpn10) to form a large central cavity in which ATP-dependent protein folding takes place. TRiC/CCT, a eukaryotic relative of Hsp60, is expressed in the cytosol and participates in the folding of actin and tubulin substrates, but lacks any association with an Hsp10-like co-factor.
- For Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse, rat, bovine, bacteria, beluga, dog, chicken, Drosophila, fish, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus HSP60. Detects a band of ~60kDa by Western blot.
Type: Primary
Antigen: HSP60
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 95046-240
- 95043-820
- 95043-818
- Antigen Symbol:
- HSP60
- HSP60
- HSP60
- Antigen Name:
- Heat Shock Protein 60
- Heat Shock Protein 60
- Heat Shock Protein 60
- Antigen Synonyms:
- Mytonin Chaperone 60|GROEL|HSP65|P60 lymphocyte protein|HLD4|HSP-60|HuCHA60|mitochondrial matrix protein P1|chaperonin 60|HSP60|mitochondrial|CPN60|short heat shock protein 60 Hsp60s1|heat shock protein 65|60 kDa chaperonin|60 kDa heat shock protein|SPG13
- Mytonin Chaperone 60|GROEL|HSP65|P60 lymphocyte protein|HLD4|HSP-60|HuCHA60|mitochondrial matrix protein P1|chaperonin 60|HSP60|mitochondrial|CPN60|short heat shock protein 60 Hsp60s1|heat shock protein 65|60 kDa chaperonin|60 kDa heat shock protein|SPG13
- Mytonin Chaperone 60|GROEL|HSP65|P60 lymphocyte protein|HLD4|HSP-60|HuCHA60|mitochondrial matrix protein P1|chaperonin 60|HSP60|mitochondrial|CPN60|short heat shock protein 60 Hsp60s1|heat shock protein 65|60 kDa chaperonin|60 kDa heat shock protein|SPG13
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Human
- Human
- Host:
- Goat
- Goat
- Goat
- Western Blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- Form:
- Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
- Purification:
- Protein G affinity purified