The Hsp70 family of heat shock proteins contains multiple homologs ranging in size from 66-78 kDa, and are the eukaryotic equivalents of the bacterial DnaK. The most studied Hsp70 members include the cytosolic stress-induced Hsp70 (Hsp72), the constitutive cytosolic Hsc70 (Hsp73), and the ER-localized BiP (Grp78). Hsp70 family members contain highly conserved N-terminal ATP-ase and C-terminal protein binding domains. Binding of peptide to Hsp70 is assisted by Hsp40, and stimulates the inherent ATPase activity of Hsp70, facilitating ATP hydrolysis and enhanced peptide binding. Hsp70 nucleotide exchange and substrate binding coordinates the folding of newly synthesized proteins, the re-folding of misfolded or denatured proteins, coordinates trafficking of proteins across cellular membranes, inhibits protein aggregation, and targets the degradation of proteins via the proteasomal pathway.
- For Western Blot
Recognizes human, mouse, rat, avian, hamster, monkey, rabbit, guinea pig, bovine, chicken, sheep, chicken, fish, pig and beluga HSP70 (HSP72). Detects a band of ~70kDa by Western blot.
Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 200062-684
- 200062-682
- Antigen Symbol:
- HSP70
- HSP70
- Antigen Name:
- Heat Shock Protein 70
- Heat Shock Protein 70
- Antigen Synonyms:
- HSP70.1/HSP70.2|HSPA1|HSPA1A|HSP-70|HSPA1B|HSP70-1/HSP70-2|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|HSX70|HSP 70
- HSP70.1/HSP70.2|HSPA1|HSPA1A|HSP-70|HSPA1B|HSP70-1/HSP70-2|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|HSX70|HSP 70
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- Form:
- Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
- Purification:
- Protein G affinity purified