Boc-cys(npys)-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: N-Boc-S-(3-nitro-2-pyridylthio)-L-cysteine
CAS Number:76880-29-0
:
Formula:C₁₃H₁₇N₃O₆S₂
MDL Number:MFCD00153301
Molecular Weight:375.414 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together