N-Boc-3-benzyloxymethyl-L-histidine
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Boc-His(Bom)-OH, N-[(1,1-Dimethylethoxy)carbonyl]-3-[(phenylmethoxy)methyl]- L-histidine, Boc-His(3-Bom)-OH
CAS Number:79950-65-5
:
Formula:C₁₉H₂₅N₃O₅
MDL Number:MFCD00043133
Molecular Weight:375.42 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together