Order Entry
United States
ContactUsLinkComponent
Ammonia Ion Selective Electrode ISE Analysis Package, Hach
Ammonia Ion Selective Electrode ISE Analysis Package, Hach
Catalog # 97021-540
Supplier:  Avantor
undefined
Ammonia Ion Selective Electrode ISE Analysis Package, Hach
Catalog # 97021-540
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Temperature range:
    0...50 °C
  • Description:
    Gas sensing: glass with replaceable NH₃ sensitive membrane
  • Material:
    Sensor body, ABS
  • Environmentally Preferable:
  • Sensor Body Material:
    ABS
  • Cable Length:
    0.91 m
  • Probe type:
    Laboratory Combination ISE
  • Range:
    0.05 mg/L to 14,000 mg/L NH3-N
  • Sensor type:
    Gas sensing: glass w/ replaceable NH3 sensitive membrane
  • Filling Solution Required:
    Ammonium Chloride
  • Junction Type:
    Porous Teflon Annular Ring
  • Cat. No.:
    97021-540
  • Junction:
    Porous PTFE annular ring
  • Fill solution:
    Ammonium chloride
  • Supplier no.:
    2348700

Specifications

About this item

Ammonia Analysis Package includes sensION combination ammonia, ISE, 5-pack of replacement ammonia membrane modules, 50 ml electrode filling solution, Hach ammonia nitrogen standard solutions in both 10 mg/L and 100 mg/L, Hach ammonia ionic strength adjustor ISA Powder Pillows pack of 100, 50 ml electrode storage solution, and 50 ml beaker.

  • Everything you need for ISE measurements in one package
  • Provides fast, stable, and accurate results
  • Easy to use
  • Flexible

Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.

The ammonia ISE is compatible with Hach sensION, sensION+, and H-Series systems.

Ordering information: sensION 51927 combination ammonia ISE (5192700), 5-pack of replacement ammonia membrane modules, 50 ml electrode filling solution, both Hach ammonia nitrogen standard solutions sizes 10 mg/L and 100 mg/L, Hach ammonia ionic strength adjustor (ISA) Powder Pillows pack of 100, 50 ml electrode storage solution, 50 ml beaker, and ISE User Manual included.