Hydrogen Peroxide Test Kit, Model HYP-1, Hach
Catalog # 89231-374
Supplier: Avantor
Product available on GSA Advantage®
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Chemical method:Thiosulfate
- Description:Hydrogen Peroxide Test Kit, Model HYP-1
- Environmentally Preferable:
- Cat. No.:89231-374
- Range:1 - 10 mg/l, 0.2 - 2.0 mg/l
- Supplier no.:2291700
- No. of tests:100
Specifications
About this item
Model HYP-1 Hydrogen Peroxide Test Kit (as H₂O₂) utilizes drop count titration and contains approximately 100 tests.
- Utilizes drop count titration a simple, economical titration method
- Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.