ContactUsLinkComponent
Digital Titrator Kit, Hach
Digital Titrator Kit, Hach
  97026-256
 :  Avantor
undefined
Digital Titrator Kit, Hach
  97026-256
 :  Avantor
  • Accuracy:
    ± 1 % (Uncertainty of readings is 1 digit. Most samples require more than 100 digits)
  • Description:
    Digital Titrator Kit
  • Environmentally Preferable:
  • Weight:
    0.29 lbs. (0.132 kg)
  • Cat. No.:
    97026-256
  • Supplier no.:
    2270900
Universal Digital Titrator Kit with Digital Titrator, manual, 100-mL graduated cylinder, 125- and 250-mL Erlenmeyer flasks, demineralizer bottle, clippers, and rugged carrying case.
  • Accurate results when and where you need them
  • Fast and Flexible
  • Versatile
  • Economical
  • EndPoint easier to determine