ContactUsLinkComponent
Data Acquisition Software, Sper Scientific
Data Acquisition Software, Sper Scientific
Catalog # 97011-798
Supplier:  Sper Scientific
undefined
Data Acquisition Software, Sper Scientific
Catalog # 97011-798
Supplier:  Sper Scientific
Supplier Number:  840052
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Data Acquisition Software
  • Environmentally Preferable:
  • Cat. No.:
    97011-798
  • Supplier no.:
    840052
Specifications
About this item
Software downloads data to your PC.
  • Provides further analysis by receiving the data stream from your meter
  • Data is saved in a text format (.txt) and can easily imported into spreadsheets, databases or word processing programs
Up to 16000 sample readings with real-time clock memory. Programmable sample interval from 1 second to 3,600 seconds. Unit displayed with changing setting. User-defined maximum (High) and minimum (Low) alarm setting. Includes one CD Rom and this operation manual.
System Requirements include Win 95 or higher operating system, 486 PC or higher withat least one serial port, RS232 Cable model 840054 sold separately above.
Works with meter models 800046, 840003, 840028, 840080, 840081, 840082, 840083, and 840085.