Order Entry
United States
ContactUsLinkComponent
Triple Side Loading Glove Box Holder, TrippNT
Triple Side Loading Glove Box Holder, TrippNT
Catalog # 89408-450
Supplier:  Avantor
undefined
Triple Side Loading Glove Box Holder, TrippNT
Catalog # 89408-450
Supplier:  Avantor

Specifications

  • Color:
    White/Clear
  • Description:
    White/Clear Triple Side Loading Glove Box Holder
  • Environmentally Preferable:
  • Cat. No.:
    89408-450
  • W×H×D:
    12×15×5"
  • Weight:
    3 lbs.
  • Supplier no.:
    50438

Specifications

About this item

This White PVC and Clear Acrylic Triple Side Loading Glove Box Holder features PVC back and sides plus a clear acrylic front

Easy-side glove box loading eliminates removal of existing boxes before refilling. Holds three boxes of gloves and includes back keyholes for installation.