GIP (1-30) amide (porcine), Glucose-Dependent Insulinotropic Polypeptide (1-30) amide (porcine)
Supplier: Bachem Americas
Certificates
About this item
This GIP fragment has potent insulinotropic activity in the isolated, perfused rat pancreas but greatly reduced somatostatinotropic activity in the isolated perfused rat stomach. The site responsible for insulinotropic activity apparently lies between residues 19 and 30 of GIP.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together