Order Entry
United States
ContactUsLinkComponent
H-Leu-His-OH
H-Leu-His-OH
  G-2520.0001BA
 :  
H-Leu-His-OH
  G-2520.0001BA
 :  4002700.0001
 :  

 

  • CAS No.:
    38062-72-5
  • Source:
  • Species:
  • Size:
    1 g
  • Environmentally Preferable:
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00037854
  • Protein/Peptide Name:
    H-Leu-His-OH
  • Molecular Weight:
    268.32
  • Sequence:
    H-Leu-His-OH
  • Molecular Formula:
    C₁₂H₂₀N₄O₃
  • Cat. No.:
    G-2520.0001BA
  • Supplier no.:
    G-2520.0001