H-Ala-Ala-OMe · HCl, Bachem
Catalog # G-1160.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:41036-19-5
- Source:
- Species:
- Size:5 g
- Environmentally Preferable:
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00237711
- Protein/Peptide Name:H-Ala-Ala-OMe · HCl
- Molecular Weight:210.66
- Sequence:H-Ala-Ala-OMe · HCl
- Molecular Formula:C₇H₁₄N₂O₃ · HCl
- Cat. No.:G-1160.0005BA
- Supplier no.:G-1160.0005
Specifications
About this item
Substrate for dipeptidyl aminopeptidase I (cathepsin C).