Order Entry
United States
ContactUsLinkComponent
Electromagnetic Coil, Large Distilling Heads, Ace Glass
Electromagnetic Coil, Large Distilling Heads, Ace Glass
Catalog # 89055-678
Supplier:  Ace Glass
Small Business Enterprise
undefined
Electromagnetic Coil, Large Distilling Heads, Ace Glass
Catalog # 89055-678
Supplier:  Ace Glass
Supplier Number:  6592-10
Small Business Enterprise

Specifications

  • Description:
    Electromagnetic Coil
  • Environmentally Preferable:
  • Cat. No.:
    89055-678
  • Supplier no.:
    6592-10

Specifications

About this item

Designed to work with large distilling heads, this electromagnetic coil has a high permeability core to operate a takeoff device on standard automatic distilling heads.

It is fastened to the head via a special clamp (comes complete with clamp and wires to connect to automatic time switch)

The apparatus is 76 mm O.D. by 44 mm high; the diameter of the metal slug is 28.6 mm.

For intermittent service only; 30 seconds maximum ON, 25% maximum duty cycle.

The unit is designed for operation at 120 volt, 50/60 Hz AC maximum, or at 24 volts DC maximum.