Type 316 Stainless Steel Agitator, Ace Glass
Supplier: Ace Glass
Certificates
About this item
The type 316 stainless steel agitator has multiple fins which may be bent, with care, to different angles
The agitator is designed for use with a 10mm shaft.
Ordering information: For replacement screw sets, see 89062-182 (for use with stainless steel shafts) or 89062-184 (for use with glass shafts).
Frequently Bought Together