Clamp, Utility, 3-Prong, Ace Glass
Catalog # 89083-216
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:3-Prong Utility Clamp, Vinylized Jaws
- Environmentally Preferable:
- Cat. No.:89083-216
- Supplier no.:11062-14
Specifications
About this item
This three-prong grip clamp has jaws and holder made of zinc alloy
The clamp can be extended in length to 16mm, and firmly locked in any position by means of the provided wing-nut. It will hold objects up to 57mm in diameter and grip rods up to 19mm in diameter.
Ordering information: Comes with an extra set of fiberglass prong covers for temperatures above 100°C.