Order Entry
United States
ContactUsLinkComponent
Buffer Solution, TSS, Thermo Scientific
Buffer Solution, TSS, Thermo Scientific
Catalog # AAJ62318-EQE
Supplier:  Avantor
CAS Number:  
Buffer Solution, TSS, Thermo Scientific
Catalog # AAJ62318-EQE
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    TSS (Transformation and Storage Solution)
  • Size:
    125 ml
  • Environmentally Preferable:
  • Cat. No.:
    AAJ62318-EQE
  • pH:
    6.5
  • Storage temperature:
    RT
  • Supplier no.:
    J62318-EQE

Specifications

About this item

TSS (transformation and storage solution) buffers can be used to make chemically competent cells.

  • Contains 85% LB medium, 10% (w/v) PEG MW 8000, 5% (v/v) DMSO, 50 mM magnesium chloride
  • Fully miscible in water
  • Sterile filtered

TSS solution is used in the one-step procedure for the preparation of competent E. coli and allows transformation of cells without heat-shock. Transformation efficiencies of 106 to 108/µg DNA are typically obtained using TSS.