About this item
SOB Medium was developed by Hanahan as a nutritionally rich growth medium for preparation and transformation of competent cells. Growth of E. coli in SOB medium results in higher transformation efficiencies of plasmids.
Applications: For cultivating recombinant strains of Escherichia coli
Contains 20g/L tryptone, 5g/L yeast extract, 2.4g/L magnesium sulfate, 0.5g/L NaCl, and 0.186g/L KCl
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
Organic Building Blocks and Intermediates